Learn More
Cayman Chemical Loganin, Purity: ≥98%, CAS: 18524-94-2, 10 mg
SDP
Catalog No. 502980737
Encompass_Preferred
An iridoid glycoside with diverse biological activities; reduces hydrogen peroxide-induced apoptosis in SH-SY5Y cells from 0.25-12.5 µM; inhibits COX-1 activity (IC50 = 3.55 µM), as well as LPS-induced TNF-α formation (IC50 = 154.6 µM) in RAW 264.7 cells; reduces food intake, blood glucose levels, and serum triglyceride levels, as well as increases serum HDL levels in db/db diabetic mice at 20 and 100 mg/kg; attenuates scopolamine-induced memory deficits in the Morris water maze and passive avoidance test in mice; reduces serum and kidney AGE and MDA levels and improves renal function in a mouse model of diabetic nephropathy at 20 and 1,000 mg/kg
Catalog No. 50-298-0737 Supplier Cayman Chemical Supplier No. 1999710 MG
An iridoid glycoside with diverse biological activities; reduces hydrogen peroxide-induced apoptosis in SH-SY5Y cells from 0.25-12.5 µM; inhibits COX-1 activity (IC50 = 3.55 µM), as well as LPS-induced TNF-α formation (IC50 = 154.6 µM) in RAW 264.7 cells; reduces food intake, blood glucose levels, and serum triglyceride levels, as well as increases serum HDL levels in db/db diabetic mice at 20 and 100 mg/kg; attenuates scopolamine-induced memory deficits in the Morris water maze and passive avoidance test in mice; reduces serum and kidney AGE and MDA levels and improves renal function in a mouse model of diabetic nephropathy at 20 and 1,000 mg/kg