Learn More
Cayman Chemical Erianin, Purity: ≥98%, CAS: 95041-90-0, 10 mg
SDP
Catalog No. 502946617
Encompass_Preferred
A benzoyl compound with diverse biological activities; inhibits the growth of H460 and H1299 lung cancer cells in a calcium/calmodulin-dependent manner; reduces tumor volume in an H460 mouse xenograft model at 100 mg/kg; increases colon length and body weight, as well as decreases colonic inflammation and the number of colonic ulcers in a mouse model of DSS-induced ulcerative colitis; decreases retinal microglia accumulation, NF-kB signaling, and TNF-α mRNA expression, as well as attenuates blood-brain barrier breakdown in a mouse model of STZ-induced diabetic retinopathy at 10 mg/kg
Catalog No. 50-294-6617 Supplier Cayman Chemical Supplier No. 3251610 MG
A benzoyl compound with diverse biological activities; inhibits the growth of H460 and H1299 lung cancer cells in a calcium/calmodulin-dependent manner; reduces tumor volume in an H460 mouse xenograft model at 100 mg/kg; increases colon length and body weight, as well as decreases colonic inflammation and the number of colonic ulcers in a mouse model of DSS-induced ulcerative colitis; decreases retinal microglia accumulation, NF-kB signaling, and TNF-α mRNA expression, as well as attenuates blood-brain barrier breakdown in a mouse model of STZ-induced diabetic retinopathy at 10 mg/kg