Learn More
Cayman Chemical Plnt Grapes ErmnIn 10mg
SDP
Catalog No. 502917389
Encompass_Preferred
A methoxylated flavone with diverse biological activities; increases the levels of intracellular ROS and decreases the levels of GSH in B16/F10 mouse melanoma cells and primary mouse melanocytes at 8 and 16 µM; reduces LPS-induced increases in iNOS and COX-2 levels in RAW 264.7 mouse macrophages (IC50s = 6.7 and 1.68 µM, respectively); inhibits viral replication of poliovirus type 1 in Vero cells (ED99 = 5 UG/ml); induces melanogenesis in zebrafish larvae at 8 and 16 µM
Catalog No. 50-291-7389 Supplier Cayman Chemical Supplier No. 3753510 MG
A methoxylated flavone with diverse biological activities; increases the levels of intracellular ROS and decreases the levels of GSH in B16/F10 mouse melanoma cells and primary mouse melanocytes at 8 and 16 µM; reduces LPS-induced increases in iNOS and COX-2 levels in RAW 264.7 mouse macrophages (IC50s = 6.7 and 1.68 µM, respectively); inhibits viral replication of poliovirus type 1 in Vero cells (ED99 = 5 UG/ml); induces melanogenesis in zebrafish larvae at 8 and 16 µM