Learn More
Cayman Chemical Plnt RhodIola rosea RosIn 1mg
SDP
Catalog No. 502946473
Encompass_Preferred
A cinnamic glycoside with antidepressant and neurotropic activities; decreases immobility time in the forced swim test in rats at 0.26 mg/kg; reduces chloral hydrate-induced sleep in mice
Catalog No. 50-294-6473 Supplier Cayman Chemical Supplier No. 343831 MG
A cinnamic glycoside with antidepressant and neurotropic activities; decreases immobility time in the forced swim test in rats at 0.26 mg/kg; reduces chloral hydrate-induced sleep in mice