Learn More
Cayman Chemical Rosin, Purity: ≥98%, CAS: 85026-55-7, 5 mg
SDP
Catalog No. 502950213
Encompass_Preferred
A cinnamic glycoside with antidepressant and neurotropic activities; decreases immobility time in the forced swim test in rats at 0.26 mg/kg; reduces chloral hydrate-induced sleep in mice
Catalog No. 50-295-0213 Supplier Cayman Chemical Supplier No. 343835 MG
A cinnamic glycoside with antidepressant and neurotropic activities; decreases immobility time in the forced swim test in rats at 0.26 mg/kg; reduces chloral hydrate-induced sleep in mice