Learn More
Cayman Chemical Plant Sterol Mixture, 1 mL
SDP
Catalog No. 502988576
Encompass_Preferred
A mixture of plant sterols; intended for use as a reference standard for the detection of these plant sterols in various sample types by GC-FID, GC-MS, or LC-MS
Catalog No. 50-298-8576 Supplier Cayman Chemical Supplier No. 293641 ML
A mixture of plant sterols; intended for use as a reference standard for the detection of these plant sterols in various sample types by GC-FID, GC-MS, or LC-MS