Learn More
Cayman Chemical Plant Sterols Kit, 1 ea
SDP
Catalog No. 502937068
Encompass_Preferred
A reference standards kit for the detection of plant sterols in various sample types by TLC, LC-MS, or GC-MS
Catalog No. 50-293-7068 Supplier Cayman Chemical Supplier No. 286431 EA
A reference standards kit for the detection of plant sterols in various sample types by TLC, LC-MS, or GC-MS