Learn More
Cayman Chemical Tomatine, Purity: ≥95%, CAS: 17406-45-0, 5 mg
SDP
Catalog No. 502968846
Encompass_Preferred
A steroidal saponin with diverse biological activities; active against a panel of 19 plant pathogenic fungi (EC50s = 13-912 µM); inhibits the growth of L. amazonensis promastigotes (IC50 = 124 nM); inhibits autophagy and induces apoptosis in SKOV3 cells at 0.75, 1, and 1.5 µM; dietary administration decreases serum LDL levels and increases fecal cholesterol secretion in hamsters at 0.1 and 0.2% w/w
Catalog No. 50-296-8846 Supplier Cayman Chemical Supplier No. 160245 MG
A steroidal saponin with diverse biological activities; active against a panel of 19 plant pathogenic fungi (EC50s = 13-912 µM); inhibits the growth of L. amazonensis promastigotes (IC50 = 124 nM); inhibits autophagy and induces apoptosis in SKOV3 cells at 0.75, 1, and 1.5 µM; dietary administration decreases serum LDL levels and increases fecal cholesterol secretion in hamsters at 0.1 and 0.2% w/w