Learn More
Cayman Chemical Plantamajoside, Purity: ≥95%, CAS: 104777-68-6, 10 mg
SDP
Catalog No. 502938694
Encompass_Preferred
A phenylethanoid with diverse biological activities; scavenges DPPH radicals in a cell-free assay (IC50 = 11.8 µM); inhibits 5- and 15-LO (IC50s = 0.375 and 96 µM, respectively); decreases viability of TE-1 ESCC cells at 20-320 µg/ml, as well as inhibits LPS-induced EMT in Eca-109 and TE-1 cells and decreases phosphorylated NF-κB levels in TE-1 cells; inhibits arachidonic acid-induced mouse ear edema at 3 mg/ear
Catalog No. 50-293-8694 Supplier Cayman Chemical Supplier No. 2729610 MG
A phenylethanoid with diverse biological activities; scavenges DPPH radicals in a cell-free assay (IC50 = 11.8 µM); inhibits 5- and 15-LO (IC50s = 0.375 and 96 µM, respectively); decreases viability of TE-1 ESCC cells at 20-320 µg/ml, as well as inhibits LPS-induced EMT in Eca-109 and TE-1 cells and decreases phosphorylated NF-κB levels in TE-1 cells; inhibits arachidonic acid-induced mouse ear edema at 3 mg/ear