Learn More
U.S. Pharmacopeia Polyethylene Glycol 3350, 25322-68-3, MFCD00081839, 1g
Catalog No. NC0006454
Encompass
Molecular formula HO-[CH2OCH2]n-OH, Melting Point 129.2 - 136.4 °F (54 - 58 °C), Boiling Point > 392 °F (> 200 °C) (decomposes)
Catalog No. NC0006454 Supplier U.S. Pharmacopeia Supplier No. 1546547
Molecular formula HO-[CH2OCH2]n-OH, Melting Point 129.2 - 136.4 °F (54 - 58 °C), Boiling Point > 392 °F (> 200 °C) (decomposes)