Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Plozasiran sodium | 2379776-41-5 | 97.9% | 15618.77 (free acid) | 1 MG
SDP

Catalog No. 5000394680
Encompass_Preferred

Plozasiran sodium is an siRNA targeting APOC3 for the study of mixed hyperlipidemia.

  • siRNA targeting APOC3
  • For the study of mixed hyperlipidemia
  • Appearance: Solid
  • Color: White to off-white
  • Shipping: Room temperature in continental US; may vary elsewhere.
  • Storage: -20°C, sealed storage, away from moisture. In solvent: -80°C, 6 months; -20°C, 1 month (sealed storage, away from moisture).

Catalog No. 50-003-94680 Supplier Medchemexpress LLC Supplier No. HY164740A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Plozasiran sodium is an siRNA targeting APOC3 for the study of mixed hyperlipidemia.

  • siRNA targeting APOC3
  • For the study of mixed hyperlipidemia
  • Appearance: Solid
  • Color: White to off-white
  • Shipping: Room temperature in continental US; may vary elsewhere.
  • Storage: -20°C, sealed storage, away from moisture. In solvent: -80°C, 6 months; -20°C, 1 month (sealed storage, away from moisture).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.