Learn More
Apexbio Technology LLC PLP (139-151) 122018-58-0 5mg
SDP
Catalog No. 5000088656
Encompass_Preferred
PLP 139-151 is a peptide derived from proteolipid protein (PLP) a major component of myelin in the central nervous system It is designed to trigger the proliferation and activation of CD4 T cells specific to myelin antigens thereby initiating autoimmune responses against neural tissues PLP 139-151 exerts its biological activity primarily through the induction of T cell-mediated immune responses In experimental autoimmune encephalomyelitis (EAE) models PLP 139-151 demonstrates immunodominant activity by facilitating disease induction Based on these pharmacological properties PLP 139-151 holds research potential in studies of T cell-mediated immune events autoimmune inflammation and the evaluation of therapeutic strategies targeting immune modulation and tolerance induction in neuroimmunological diseases
Catalog No. 50-000-88656 Supplier Apexbio Technology LLC Supplier No. B53155
PLP 139-151 is a peptide derived from proteolipid protein (PLP) a major component of myelin in the central nervous system It is designed to trigger the proliferation and activation of CD4 T cells specific to myelin antigens thereby initiating autoimmune responses against neural tissues PLP 139-151 exerts its biological activity primarily through the induction of T cell-mediated immune responses In experimental autoimmune encephalomyelitis (EAE) models PLP 139-151 demonstrates immunodominant activity by facilitating disease induction Based on these pharmacological properties PLP 139-151 holds research potential in studies of T cell-mediated immune events autoimmune inflammation and the evaluation of therapeutic strategies targeting immune modulation and tolerance induction in neuroimmunological diseases