Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PLP (180-199) TFA | 99.22% | 2085.34 (free base) | 1 MG
SDP

Catalog No. 5000396991
Encompass_Preferred

PLP (180-199) TFA is the TFA salt form of PLP (180-199). It induces the activation and differentiation of T cells and induces experimental autoimmune encephalomyelitis (EAE) in mouse models.

  • Activates and differentiates T cells
  • Induces experimental autoimmune encephalomyelitis (EAE) in mouse models
  • Molecular weight: 2085.34 (free base)
  • Purity: 99.22%
  • Sealed storage, away from moisture
  • Powder storage: -80°C for 2 years, -20°C for 1 year
  • In solvent storage: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-003-96991 Supplier Medchemexpress LLC Supplier No. HYP5462A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PLP (180-199) TFA is the TFA salt form of PLP (180-199). It induces the activation and differentiation of T cells and induces experimental autoimmune encephalomyelitis (EAE) in mouse models.

  • Activates and differentiates T cells
  • Induces experimental autoimmune encephalomyelitis (EAE) in mouse models
  • Molecular weight: 2085.34 (free base)
  • Purity: 99.22%
  • Sealed storage, away from moisture
  • Powder storage: -80°C for 2 years, -20°C for 1 year
  • In solvent storage: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.