Learn More
Aviva Systems Biology Corp PLP1 Antibody - N-terminal region (ARP33843_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33843_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PLP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PLP1. Alternate names: MMPL, PLP, PLP/DM20, PMD, SPG2, HLD1ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55168