Learn More
Cayman Chemical PLP139-151trIfluoroaceta 10mg
SDP
Supplier:  Cayman Chemical 3315910 MG
Encompass_Preferred
An encephalitogenic peptide; induces the formation of inflammatory foci, axonal injury, and demyelination in the spinal cord, as well as motor deficits in mice at 150 UG/animal; commonly used to induce EAE in mice
Catalog No. 50-294-6549