Learn More
Aviva Systems Biology Corp PLP2 Antibody - middle region (ARP45348_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP45348_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PLP2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PLP2. Alternate names: A4, A4-LSB, MGC126187, A4LSBApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Sheep
Catalog No. 50-014-53446