Learn More
Medchemexpress LLC PLpro inhibitor | 1093070-14-4 | 99.9% | 346.42 | 10 MM 1 ML
SDP
Catalog No. 5000386491
Encompass_Preferred
PLpro inhibitor is a potent inhibitor of papain-like protease (PLpro) with an IC50 of 2.6 μM. It inhibits SARS-CoV-2 PLpro with an IC50 of 5.0 μM and an EC50 of 21.0 μM. It is a noncovalent inhibitor that blocks SARS virus replication and shows significant antiviral activity with EC50 values of 13.1 μM without toxicity up to the highest concentration tested. The increasing antiviral potency correlates with the in vitro inhibition of PLpro, suggesting a direct action on the enzyme in cells.
  • Potent inhibitor of papain-like protease (PLpro) (IC50 = 2.6 μM).
  • Inhibits SARS-CoV-2 PLpro (IC50 = 5.0 μM, EC50 = 21.0 μM).
  • Displays significant antiviral activity (EC50 = 13.1 μM) without toxicity.
  • Noncovalent class of inhibitor that blocks SARS virus replication.
Catalog No. 50-003-86491 Supplier Medchemexpress LLC Supplier No. HY1754210MM/1ML
PLpro inhibitor is a potent inhibitor of papain-like protease (PLpro) with an IC50 of 2.6 μM. It inhibits SARS-CoV-2 PLpro with an IC50 of 5.0 μM and an EC50 of 21.0 μM. It is a noncovalent inhibitor that blocks SARS virus replication and shows significant antiviral activity with EC50 values of 13.1 μM without toxicity up to the highest concentration tested. The increasing antiviral potency correlates with the in vitro inhibition of PLpro, suggesting a direct action on the enzyme in cells.
  • Potent inhibitor of papain-like protease (PLpro) (IC50 = 2.6 μM).
  • Inhibits SARS-CoV-2 PLpro (IC50 = 5.0 μM, EC50 = 21.0 μM).
  • Displays significant antiviral activity (EC50 = 13.1 μM) without toxicity.
  • Noncovalent class of inhibitor that blocks SARS virus replication.