Learn More
Ubpbio Containing recombinant papain-like protease (PLPro) encoded by SARS-CoV-2 virus, fluorogenic substrate for its proteinase activity, control inhibitor, and buffer optimized for room temperature assay operation. 1,200 assays included
Catalog No. 502339256
Encompass_Preferred
The kit measures the proteinase activity of SARS-CoV-2 papain-like protease (PLPro), can be used for screening/profiling PLPro inhibitors. Kit I is sufficient for 120 X 50µl reactions; Kit II is sufficient for 1,200 X 50µl reactions. Optimized for rapidly and quantitatively measuring PLPro activity under room temperature.
Catalog No. 50-233-9256 Supplier Ubpbio Supplier No. J7611
The kit measures the proteinase activity of SARS-CoV-2 papain-like protease (PLPro), can be used for screening/profiling PLPro inhibitors. Kit I is sufficient for 120 X 50µl reactions; Kit II is sufficient for 1,200 X 50µl reactions. Optimized for rapidly and quantitatively measuring PLPro activity under room temperature.