Learn More
MSC Partners Brand Polystyrene Foam Insert, For 1 gal. Jug, 48/Pack
Catalog No. 501919827
Encompass
These foam inserts are designed to protect 1-gallon plastic jugs during shipping and storage. Made from expanded polystyrene, they help prevent damage to canisters and spillage of contents by providing cushioning and stability.
  • Manufactured from expanded polystyrene with a nominal density of 0.75 lb per cubic foot
  • Dimensions: 6 in. wide, 6 in. long, 1 in. high
  • Color: White
  • Ensures protection for industry-standard size containers
  • Shippers are responsible for correct packaging and final closure per CFR 49 178.601B
Catalog No. 50-191-9827 Supplier MSC Supplier No. 39594460
These foam inserts are designed to protect 1-gallon plastic jugs during shipping and storage. Made from expanded polystyrene, they help prevent damage to canisters and spillage of contents by providing cushioning and stability.
  • Manufactured from expanded polystyrene with a nominal density of 0.75 lb per cubic foot
  • Dimensions: 6 in. wide, 6 in. long, 1 in. high
  • Color: White
  • Ensures protection for industry-standard size containers
  • Shippers are responsible for correct packaging and final closure per CFR 49 178.601B