Learn More
MSC Partners Brand Strap Guards, 2 1/2 in, 1000/Pack
Catalog No. 501917890
Encompass
Plastic strap guards are designed to protect palletized products from strapping damage during shipping and warehousing. These durable plastic guards help stabilize loads, improve stacking strength, and prevent shifting by distributing strapping tension effectively.
  • Prevents damage during shipping and warehousing
  • Stabilizes loads and improves stacking strength
  • Provides additional support for double stacking pallets
  • Eliminates shifting loads
  • Protects palletized products from strapping damage
  • Spreads strapping tension to prevent damage
  • Ribbed channel stops strapping from moving
  • Can be used with steel or poly strapping
  • Reusable and recyclable
  • Constructed from strong, durable plastic
Catalog No. 50-191-7890 Supplier MSC Supplier No. 39574744
Plastic strap guards are designed to protect palletized products from strapping damage during shipping and warehousing. These durable plastic guards help stabilize loads, improve stacking strength, and prevent shifting by distributing strapping tension effectively.
  • Prevents damage during shipping and warehousing
  • Stabilizes loads and improves stacking strength
  • Provides additional support for double stacking pallets
  • Eliminates shifting loads
  • Protects palletized products from strapping damage
  • Spreads strapping tension to prevent damage
  • Ribbed channel stops strapping from moving
  • Can be used with steel or poly strapping
  • Reusable and recyclable
  • Constructed from strong, durable plastic