Learn More
ABclonal Technology PLS1 Rabbit pAb
SDP
Catalog No. 5000105252
Encompass_Preferred
Plastins are a family of actin-binding proteins that are conserved throughout eukaryote evolution and expressed in most tissues of higher eukaryotes. In humans, two ubiquitous plastin isoforms (L and T) have been identified. The protein encoded by this gene is a third distinct plastin isoform, which is specifically expressed at high levels in the small intestine. Alternatively spliced transcript variants varying in the 5 UTR, but encoding the same protein, have been found for this gene. A pseudogene of this gene is found on chromosome 11.
Catalog No. 50-001-05252 Supplier ABclonal Technology Supplier No. A15303500UL
Plastins are a family of actin-binding proteins that are conserved throughout eukaryote evolution and expressed in most tissues of higher eukaryotes. In humans, two ubiquitous plastin isoforms (L and T) have been identified. The protein encoded by this gene is a third distinct plastin isoform, which is specifically expressed at high levels in the small intestine. Alternatively spliced transcript variants varying in the 5 UTR, but encoding the same protein, have been found for this gene. A pseudogene of this gene is found on chromosome 11.