Learn More
Aviva Systems Biology Corp PLS3 Antibody - middle region (ARP56623_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56623_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PLS3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PLS3. Alternate names: t-plastin, T-plastinApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast
Catalog No. 50-014-61928