Learn More
Aviva Systems Biology Corp PLSCR1 Antibody - N-terminal region (ARP57513_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57513_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PLSCR1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PLSCR1. Alternate names: MMTRA1BApplicationWB, IHC. Species reactivity: Human
Catalog No. 50-014-53885