Learn More
Innovative Research Inc Plsmin Chromo Sbstrt Lyo 25mg
Catalog No. 502744631
Encompass_Preferred
Thrombin Chromogenic Substrate Lyophillized 25mg - Thrombin Chromogenic Substrate Lyophillized from Innovative Research has been synthesized to be chemically and functionally identical to Chromogenix S-2238. This is a Lyophilized powder buffered in mannitol and a purity of 98% by HPLC analysis. This product is useful for for chromogenic determination of thrombin activity. For best results add 4.0 ml of 1mM HCl directly to vial to make a 10mM stock solution then aliquot and freeze. Dilute 150 in PBS or TBS to make an 0.2mM working solution. Add 0.1 ml test sample to 0.9 ml substrate and monitor color development at 405nm. Molecular Weight 625.6 Da. Formula H-D-Phe-Pip-Arg-pNA . 2HCl.
Catalog No. 50-274-4631 Supplier Innovative Research Inc Supplier No. IAFTHRCGSLY25MG
Thrombin Chromogenic Substrate Lyophillized 25mg - Thrombin Chromogenic Substrate Lyophillized from Innovative Research has been synthesized to be chemically and functionally identical to Chromogenix S-2238. This is a Lyophilized powder buffered in mannitol and a purity of 98% by HPLC analysis. This product is useful for for chromogenic determination of thrombin activity. For best results add 4.0 ml of 1mM HCl directly to vial to make a 10mM stock solution then aliquot and freeze. Dilute 150 in PBS or TBS to make an 0.2mM working solution. Add 0.1 ml test sample to 0.9 ml substrate and monitor color development at 405nm. Molecular Weight 625.6 Da. Formula H-D-Phe-Pip-Arg-pNA . 2HCl.