Learn More
MSC PFERD Guide, Plastic, For Plastic Handle, 25-30 Degree Angle
Catalog No. 502838515
Encompass
This plastic guide is designed for use with a file handle, specifically for manual chain saw filing applications. It aids in achieving consistent sharpening results by maintaining a precise 25-30 degree sharpening angle. This type of guide is often provided as a component within a comprehensive file handle and sharpening gauge set.
  • Supports consistent sharpening results
  • Provides a precise 25-30 degree sharpening angle
  • Designed for attachment to a file handle
  • Suitable for manual chain saw filing
  • Constructed from durable plastic
Catalog No. 50-283-8515 Supplier MSC Supplier No. 49175722
This plastic guide is designed for use with a file handle, specifically for manual chain saw filing applications. It aids in achieving consistent sharpening results by maintaining a precise 25-30 degree sharpening angle. This type of guide is often provided as a component within a comprehensive file handle and sharpening gauge set.
  • Supports consistent sharpening results
  • Provides a precise 25-30 degree sharpening angle
  • Designed for attachment to a file handle
  • Suitable for manual chain saw filing
  • Constructed from durable plastic