Learn More
MSC PFERD Handle Guide, Plastic, 30-35 Degree Angle
Catalog No. 502838514
Encompass
This product provides guides for plastic handles, specifically designed to assist in sharpening chainsaw files. These guides ensure consistent sharpening results by maintaining the correct sharpening angle, making them suitable for use with an ergonomic file handle.
  • Provides a sharpening angle of 30 to 35 degrees.
  • Supports consistent sharpening results.
  • Attaches to the stud of an ergonomic file handle.
Catalog No. 50-283-8514 Supplier MSC Supplier No. 49175714
This product provides guides for plastic handles, specifically designed to assist in sharpening chainsaw files. These guides ensure consistent sharpening results by maintaining the correct sharpening angle, making them suitable for use with an ergonomic file handle.
  • Provides a sharpening angle of 30 to 35 degrees.
  • Supports consistent sharpening results.
  • Attaches to the stud of an ergonomic file handle.