Learn More
MSC Collomix Mixer Cleaning Bucket, 8 gal, Brushes, 1 ea
Catalog No. 502818291
Encompass
This cleaning bucket is designed for quickly and easily cleaning mixing paddles. It helps remove residual material from mixing paddles, extending their lifespan and ensuring optimal performance.
  • Effective bucket and brush system
  • Cleans mixing paddles rapidly
  • Extends paddle lifespan
  • Ensures proper functionality of paddles
  • Removable lid for convenient emptying
  • Rugged 8-gallon capacity
  • Integrated carrying handles
Catalog No. 50-281-8291 Supplier MSC Supplier No. 11608122
This cleaning bucket is designed for quickly and easily cleaning mixing paddles. It helps remove residual material from mixing paddles, extending their lifespan and ensuring optimal performance.
  • Effective bucket and brush system
  • Cleans mixing paddles rapidly
  • Extends paddle lifespan
  • Ensures proper functionality of paddles
  • Removable lid for convenient emptying
  • Rugged 8-gallon capacity
  • Integrated carrying handles