Learn More
Grainger DuraShelf Plastic Shelving, Solid Top, HDPE, 2-Shelf System, 1 ea
Catalog No. 502751811
Encompass_Preferred
This DuraShelf® 2-shelf system is a durable, industrial-grade storage solution constructed from high-density polyethylene (HDPE). Designed for versatility in indoor and outdoor environments, this shelving unit is UV-treated, corrosion-resistant, and suitable for chemical storage. Its solid shelf design prevents cross-product contamination, while the modular, no-tool assembly ensures quick and efficient setup.
  • Constructed from durable, chemically resistant HDPE
  • Solid shelf design prevents cross-product contamination
  • Features UV-treated material suitable for indoor or outdoor use
  • Provides a total load capacity of 660 lb (330 lb per shelf)
  • Simple no-tool assembly process
  • Overall dimensions are 36 in. W x 24 in. D x 27 in. H
Catalog No. 50-275-1811 Supplier Grainger Supplier No. 14U077
This DuraShelf® 2-shelf system is a durable, industrial-grade storage solution constructed from high-density polyethylene (HDPE). Designed for versatility in indoor and outdoor environments, this shelving unit is UV-treated, corrosion-resistant, and suitable for chemical storage. Its solid shelf design prevents cross-product contamination, while the modular, no-tool assembly ensures quick and efficient setup.
  • Constructed from durable, chemically resistant HDPE
  • Solid shelf design prevents cross-product contamination
  • Features UV-treated material suitable for indoor or outdoor use
  • Provides a total load capacity of 660 lb (330 lb per shelf)
  • Simple no-tool assembly process
  • Overall dimensions are 36 in. W x 24 in. D x 27 in. H