Learn More
Grainger Durashelf Plastic Shelving, 96 in x 24 in, 27 in H, 2-Shelf System, 1 ea
Catalog No. 502752461
Encompass_Preferred
This freestanding plastic shelving system provides a durable storage solution for commercial, industrial, or maintenance environments. Constructed from high-density polyethylene (HDPE) with solid-top shelves, this unit features excellent corrosion and chemical resistance, making it suitable for specialized storage needs including hazardous materials. The UV-treated construction allows for both indoor and outdoor use. The two-shelf system arrives unassembled and is designed for easy, no-tool assembly.
  • Features a commercial strength rating with an 885 lb. load capacity per shelf and a 1,770 lb. total unit capacity.
  • Provides a 96 in. (width) x 24 in. (depth) x 27 in. (height) footprint with 21-3/8 in. of vertical shelf spacing.
  • Includes eight leg connectors and eight posts.
  • UV treated for use in diverse indoor or outdoor settings.
Catalog No. 50-275-2461 Supplier Grainger Supplier No. 14U082
This freestanding plastic shelving system provides a durable storage solution for commercial, industrial, or maintenance environments. Constructed from high-density polyethylene (HDPE) with solid-top shelves, this unit features excellent corrosion and chemical resistance, making it suitable for specialized storage needs including hazardous materials. The UV-treated construction allows for both indoor and outdoor use. The two-shelf system arrives unassembled and is designed for easy, no-tool assembly.
  • Features a commercial strength rating with an 885 lb. load capacity per shelf and a 1,770 lb. total unit capacity.
  • Provides a 96 in. (width) x 24 in. (depth) x 27 in. (height) footprint with 21-3/8 in. of vertical shelf spacing.
  • Includes eight leg connectors and eight posts.
  • UV treated for use in diverse indoor or outdoor settings.