Learn More
Grainger DuraShelf® Freestanding Plastic Shelving, HDPE, 3-Shelf, 36 in W x 24 in D x 51 in H
Catalog No. 502750124
Encompass_Preferred
This freestanding open-style shelving unit provides a durable and versatile storage solution suitable for a wide range of industrial, indoor, or outdoor applications. Constructed from high-density polyethylene, the unit offers excellent corrosion resistance and supports a total load capacity of 990 lb.
  • Features three solid shelves made of high-density polyethylene
  • Supports 330 lb per individual shelf
  • Includes 8 leg connectors and 8 posts for assembly
  • Offers fixed shelf adjustability for stable configuration
  • Corrosion-resistant material suitable for varied environments
Catalog No. 50-275-0124 Supplier Grainger Supplier No. 14U084
This freestanding open-style shelving unit provides a durable and versatile storage solution suitable for a wide range of industrial, indoor, or outdoor applications. Constructed from high-density polyethylene, the unit offers excellent corrosion resistance and supports a total load capacity of 990 lb.
  • Features three solid shelves made of high-density polyethylene
  • Supports 330 lb per individual shelf
  • Includes 8 leg connectors and 8 posts for assembly
  • Offers fixed shelf adjustability for stable configuration
  • Corrosion-resistant material suitable for varied environments