Learn More
Grainger Durashelf® Plastic Shelving, 3-Shelf, 66 in x 24 in, 1 ea.
Catalog No. 502753051
Encompass_Preferred
This freestanding all-plastic storage unit is designed for demanding industrial and commercial environments, providing a robust solution for organizing essential inventory and supplies. Engineered from high-density polyethylene, these units are suitable for storing diverse materials, including chemicals and food containers, while maintaining structural integrity in dusty or challenging settings.
  • Features three high-density polyethylene shelves with a 605 lb. capacity each.
  • Offers a total load capacity of 1,820 lb.
  • Promotes air circulation and limits debris accumulation with a flow-through grid design.
  • Corrosion-resistant, chemical-resistant, and UV-treated for long-term durability.
  • Enables tool-free assembly and includes components for linking additional units.
  • Made in the USA.
Catalog No. 50-275-3051 Supplier Grainger Supplier No. 14U087
This freestanding all-plastic storage unit is designed for demanding industrial and commercial environments, providing a robust solution for organizing essential inventory and supplies. Engineered from high-density polyethylene, these units are suitable for storing diverse materials, including chemicals and food containers, while maintaining structural integrity in dusty or challenging settings.
  • Features three high-density polyethylene shelves with a 605 lb. capacity each.
  • Offers a total load capacity of 1,820 lb.
  • Promotes air circulation and limits debris accumulation with a flow-through grid design.
  • Corrosion-resistant, chemical-resistant, and UV-treated for long-term durability.
  • Enables tool-free assembly and includes components for linking additional units.
  • Made in the USA.