Learn More
Grainger Durashelf® Plastic Shelving, HDPE, 48 in x 24 in, 4-Shelf System
Catalog No. 502752450
Encompass_Preferred
This durable, freestanding plastic shelving unit is constructed from high-density polyethylene (HDPE) and features a solid-top design. It is engineered for versatility and longevity, suitable for both indoor and outdoor applications in environments such as HAZMAT storage and maintenance rooms.
  • Features corrosion-resistant and rust-proof properties
  • Includes four 48 in. x 24 in. solid shelves
  • Offers a total load capacity of 1,760 lb.
  • Utilizes a no-tool assembly design
  • Provides UV-treated surfaces for enhanced durability
  • Adjustable in 4 in. increments
  • Supports use for preventing cross-product contamination
Catalog No. 50-275-2450 Supplier Grainger Supplier No. 14U093
This durable, freestanding plastic shelving unit is constructed from high-density polyethylene (HDPE) and features a solid-top design. It is engineered for versatility and longevity, suitable for both indoor and outdoor applications in environments such as HAZMAT storage and maintenance rooms.
  • Features corrosion-resistant and rust-proof properties
  • Includes four 48 in. x 24 in. solid shelves
  • Offers a total load capacity of 1,760 lb.
  • Utilizes a no-tool assembly design
  • Provides UV-treated surfaces for enhanced durability
  • Adjustable in 4 in. increments
  • Supports use for preventing cross-product contamination