Learn More
Grainger Durashelf Plastic Shelving, HDPE, 66 in W, 4-Shelf System, 1 ea
Catalog No. 502752008
Encompass_Preferred
This freestanding four-shelf bulk storage rack is constructed from high-density polyethylene (HDPE) to provide a durable and corrosion-resistant solution for various storage environments. Featuring a solid-top design and molded construction, this system is suitable for indoor and outdoor use and supports demanding storage requirements with a total load capacity of 2,420 lb.
  • Features corrosion-resistant and chemically resistant material properties
  • Supports heavy-duty applications with a 605 lb capacity per shelf
  • Designed for no-tool assembly for efficient setup
  • Includes UV-treated components for enhanced durability
  • Constructed from recycled materials
  • Ideal for hazardous material storage and applications requiring the prevention of cross-product contamination
Catalog No. 50-275-2008 Supplier Grainger Supplier No. 14U094
This freestanding four-shelf bulk storage rack is constructed from high-density polyethylene (HDPE) to provide a durable and corrosion-resistant solution for various storage environments. Featuring a solid-top design and molded construction, this system is suitable for indoor and outdoor use and supports demanding storage requirements with a total load capacity of 2,420 lb.
  • Features corrosion-resistant and chemically resistant material properties
  • Supports heavy-duty applications with a 605 lb capacity per shelf
  • Designed for no-tool assembly for efficient setup
  • Includes UV-treated components for enhanced durability
  • Constructed from recycled materials
  • Ideal for hazardous material storage and applications requiring the prevention of cross-product contamination