Learn More
Grainger Plstc Shelf 36inx51inx36inShlfLdCp 490lb
Catalog No. 502748741
Encompass_Preferred
The Grainger 14U085 is a 14-inch diameter, 85 CFM, 115V, 1/6 HP exhaust fan. It features a galvanized steel housing with baked enamel finish, thermally protected motor, permanently lubricated sleeve bearings, and includes mounting hardware. This exhaust fan is suitable for ventilating bathrooms, laundry rooms, utility rooms and other household applications. With a noise rating of 6.5 sones, it provides efficient airflow while maintaining relatively quiet operation. The maximum static pressure is 0.25 in. w.g. and it draws 1.1 amps. This UL/cUL listed product comes with a 1-year warranty from Grainger.
Catalog No. 50-274-8741 Supplier Grainger Supplier No. 14U085
The Grainger 14U085 is a 14-inch diameter, 85 CFM, 115V, 1/6 HP exhaust fan. It features a galvanized steel housing with baked enamel finish, thermally protected motor, permanently lubricated sleeve bearings, and includes mounting hardware. This exhaust fan is suitable for ventilating bathrooms, laundry rooms, utility rooms and other household applications. With a noise rating of 6.5 sones, it provides efficient airflow while maintaining relatively quiet operation. The maximum static pressure is 0.25 in. w.g. and it draws 1.1 amps. This UL/cUL listed product comes with a 1-year warranty from Grainger.