Learn More
Grainger DuraShelf® Plastic Shelving, 66 in. x 36 in., 4-Shelf, 3,600 lbs. Capacity, 1 ea.
Catalog No. 502748764
Encompass_Preferred
These stationary high-density polyethylene shelving units provide reliable storage for industrial environments, including HAZMAT storage and maintenance rooms. Designed for high durability, these units are resistant to chemicals, abrasion, and impacts, and they remain impervious to oils, salts, and acids. Featuring a fixed-shelf design, the unit offers a total load capacity of 3,600 lb, with each of the four solid shelves supporting up to 895 lb.
  • Constructed from high-density polyethylene for chemical resistance and durability.
  • Fixed shelf design with no-tool, press-fit assembly.
  • Impervious to oils, salts, acids, and most chemicals.
  • Steam cleanable and maintenance-free design.
  • Suitable for both indoor and outdoor use.
  • UV treated for extended service life.
  • Will not rust or corrode.
Catalog No. 50-274-8764 Supplier Grainger Supplier No. 14U095
These stationary high-density polyethylene shelving units provide reliable storage for industrial environments, including HAZMAT storage and maintenance rooms. Designed for high durability, these units are resistant to chemicals, abrasion, and impacts, and they remain impervious to oils, salts, and acids. Featuring a fixed-shelf design, the unit offers a total load capacity of 3,600 lb, with each of the four solid shelves supporting up to 895 lb.
  • Constructed from high-density polyethylene for chemical resistance and durability.
  • Fixed shelf design with no-tool, press-fit assembly.
  • Impervious to oils, salts, acids, and most chemicals.
  • Steam cleanable and maintenance-free design.
  • Suitable for both indoor and outdoor use.
  • UV treated for extended service life.
  • Will not rust or corrode.