Learn More
Electron Microscopy Sciences CytoSep™ Plastic Cytology Single Funnel Tubing with Brown Filter & Cap
SDP
Catalog No. 501900459
Encompass_Preferred
CytoSep Cytology Funnels are totally disposable, eliminating time consuming decontamination and cleaning. They are safe to use and reduce the risk of exposure to pathologic samples.They also decrease the possibility of cross-contamination.
This series of CytoSep Funnels is single-use. The disposable sample chambers do not need metal clips and are designed to meet any processing equipment. They are capable producing high quality thin-layer slide preparation while improving laboratory efficiency. Brown Filter Cards for sample volumes up to 0.4 ml.
Catalog No. 50-190-0459 Supplier Electron Microscopy Sciences Supplier No. 6275220
CytoSep Cytology Funnels are totally disposable, eliminating time consuming decontamination and cleaning. They are safe to use and reduce the risk of exposure to pathologic samples.They also decrease the possibility of cross-contamination.
This series of CytoSep Funnels is single-use. The disposable sample chambers do not need metal clips and are designed to meet any processing equipment. They are capable producing high quality thin-layer slide preparation while improving laboratory efficiency. Brown Filter Cards for sample volumes up to 0.4 ml.