Learn More
Fine Science Tools Plastic Sterilization Containers, 25 x 15 x 2 cm
SDP
Catalog No. NC1466520
Encompass
Plastic Sterilization Containers, Single Tier, 25 x 15 x 2cmThis sterilization container is made from Ultem polymer to ensure strength and long lasting durability. The transparent perforated lid provides easy identification of your surgical instruments. Silicone mat is included for a safe and secure hold. Trays are also stackable for convenience. Suitable for autoclave or ethylene oxide sterilization.
Catalog No. NC1466520 Supplier Fine Science Tools Supplier No. 2084003
Plastic Sterilization Containers, Single Tier, 25 x 15 x 2cmThis sterilization container is made from Ultem polymer to ensure strength and long lasting durability. The transparent perforated lid provides easy identification of your surgical instruments. Silicone mat is included for a safe and secure hold. Trays are also stackable for convenience. Suitable for autoclave or ethylene oxide sterilization.