Learn More
Electron Microscopy Sciences Plastic Weighing Boat Medium, 3-5/16" x ¾"H
SDP
Catalog No. 501899211
Encompass_Preferred
They are utilized to weigh liquid or solid samples. Anti-static. Recessed corners facilitate controlled pour-off. Square shape, contour bottom.
Size: 3-5/16" x ¾"H
Catalog No. 50-189-9211 Supplier Electron Microscopy Sciences Supplier No. 70041
They are utilized to weigh liquid or solid samples. Anti-static. Recessed corners facilitate controlled pour-off. Square shape, contour bottom.
Size: 3-5/16" x ¾"H