Learn More
Grainger Structural Plastics Plastic Shelving, 96 in x 36 in, 51 in H, 3 Shelves, 1 ea
Catalog No. 502752599
Encompass_Preferred
These stationary plastic shelving units provide a high-durability, commercial-grade storage solution designed to resist chemicals, abrasion, and impacts. Featuring a press-fit assembly design, these units are easily constructed without tools by pressing components together for a secure connection. The system includes three solid-style shelves with a grid top, offering a robust, fixed-shelf configuration suitable for various demanding environments.
  • Overall dimensions of 96 in. W x 36 in. D x 51 in. H
  • Constructed from high density polyethylene for enhanced durability
  • Whole unit load capacity of 3,960 lb
  • Load capacity of 1,320 lb per shelf
  • Fixed shelf spacing of 21-3/8 in. apart
  • Corrosion-resistant material properties
  • Black finish
Catalog No. 50-275-2599 Supplier Grainger Supplier No. 14U090
These stationary plastic shelving units provide a high-durability, commercial-grade storage solution designed to resist chemicals, abrasion, and impacts. Featuring a press-fit assembly design, these units are easily constructed without tools by pressing components together for a secure connection. The system includes three solid-style shelves with a grid top, offering a robust, fixed-shelf configuration suitable for various demanding environments.
  • Overall dimensions of 96 in. W x 36 in. D x 51 in. H
  • Constructed from high density polyethylene for enhanced durability
  • Whole unit load capacity of 3,960 lb
  • Load capacity of 1,320 lb per shelf
  • Fixed shelf spacing of 21-3/8 in. apart
  • Corrosion-resistant material properties
  • Black finish