Learn More
Uline Plastic Bottles with Sprayers - 16 oz, Translucent white, 6 EA
Catalog No. NC2199641
Encompass
Tough plastic bottles marked with ounce and milliliter measurements, Standard blue nozzle included, Sprayers adjust from mist to steady stream, Graduation markings range from 1 to 12 ounces, The fill ring at the top of the neck indicates the full 16 ounce capacity, Standard nozzles are not leakproof, BPA free, Not FDA Compliant, Not chemical resistant
This item is not returnable. View return policy
Catalog No. NC2199641 Supplier Uline Supplier No. S11686CS
Tough plastic bottles marked with ounce and milliliter measurements, Standard blue nozzle included, Sprayers adjust from mist to steady stream, Graduation markings range from 1 to 12 ounces, The fill ring at the top of the neck indicates the full 16 ounce capacity, Standard nozzles are not leakproof, BPA free, Not FDA Compliant, Not chemical resistant