Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Analytical Sales & Services Plate for 13mm Test Tubes

Catalog No. 50143024
Encompass

Advantage Plate for 13mmTest Tubes for 12 Position Vacuum Manifold

Catalog No. 50-143-024 Supplier Analytical Sales & Services Supplier No. 210612
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Advantage Plate for 13mmTest Tubes for 12 Position Vacuum Manifold

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.