Learn More
MSC MSC Pallet Cover, 46x42x68 in, 2 mil, 50 Pack
Catalog No. 501919796
Encompass
These black pallet cover liners are designed to protect equipment and pallets from dirt, dust, and moisture. With a 2 mil thickness, they are suitable for various protective and concealment applications, including lining large cartons.
  • Large, gusseted poly bags
  • Opaque black material conceals contents and prevents pilferage
  • Protects items from dirt, dust, and moisture
  • Pre-cut and perforated on a roll for easy dispensing
  • Ideal for outside storage
Catalog No. 50-191-9796 Supplier MSC Supplier No. 39594148
These black pallet cover liners are designed to protect equipment and pallets from dirt, dust, and moisture. With a 2 mil thickness, they are suitable for various protective and concealment applications, including lining large cartons.
  • Large, gusseted poly bags
  • Opaque black material conceals contents and prevents pilferage
  • Protects items from dirt, dust, and moisture
  • Pre-cut and perforated on a roll for easy dispensing
  • Ideal for outside storage