Learn More
Edge Biosystems Inc Adhesive Plate Sealers, 100 per pk
SDP
Catalog No. NC9754448
Encompass
Our Adhesive Plate Sealers can be used for protecting samples in 96-well and 384-well plates during shaking, incubation, and storage.They are packaged in sets of 25 sealers and can be secured to any flat surface to eliminate waste and clutter.Adhesive Plate Sealers are resistant to heat and cold (-20C to 100C).
Catalog No. NC9754448 Supplier Edge Biosystems Inc Supplier No. 48461
Our Adhesive Plate Sealers can be used for protecting samples in 96-well and 384-well plates during shaking, incubation, and storage.They are packaged in sets of 25 sealers and can be secured to any flat surface to eliminate waste and clutter.Adhesive Plate Sealers are resistant to heat and cold (-20C to 100C).