Learn More
GenDEPOT amfiRivert Platinum One cDNA Synthesis Master Mix(4X) 50rxns
Catalog No. NC2751401
Encompass_Preferred
amfiRivert cDNA One cDNA Synthesis Platinum Master Mix (4X) is specially designed for high-yield Reverse transcription system that all provides components needed for cDNA synthesis. The master mix produces excellent results in both real-time and conventional RT-PCR. amfiRivert Platinum One cDNA Master Mix (4X) is fully active at 45-55 oC, providing the higher cDNA yields and successful cDNA synthesis for diffucult RNA transcription. amfiRivert Platinum One cDNA Synthesis Master Mix (4X) includes an optimized primer mix which results in the generation of first -strand cDNAs from an entire transcript without the end-bias observed with typical oligo (dT)n or random hexamer primers. This mixed primer strategy over-comes variability in real-time PCR gene expression analysis that can result from using different individual primers.
Catalog No. NC2751401 Supplier GenDEPOT Supplier No. R6100050
amfiRivert cDNA One cDNA Synthesis Platinum Master Mix (4X) is specially designed for high-yield Reverse transcription system that all provides components needed for cDNA synthesis. The master mix produces excellent results in both real-time and conventional RT-PCR. amfiRivert Platinum One cDNA Master Mix (4X) is fully active at 45-55 oC, providing the higher cDNA yields and successful cDNA synthesis for diffucult RNA transcription. amfiRivert Platinum One cDNA Synthesis Master Mix (4X) includes an optimized primer mix which results in the generation of first -strand cDNAs from an entire transcript without the end-bias observed with typical oligo (dT)n or random hexamer primers. This mixed primer strategy over-comes variability in real-time PCR gene expression analysis that can result from using different individual primers.