Learn More
Apexbio Technology LLC PLTP-Act-Fluoro-Assay-Kit
SDP
Catalog No. 5000567796
Encompass_Preferred
PLTP Activity Fluorometric Assay Kit is utilized in biomedical research to quantify phospholipid transfer protein (PLTP) activity This kit is often used in studies focused on lipid metabolism and cardiovascular disease The assay operates through the detection of fluorescent signals generated when PLTP catalyzes the transfer of phospholipids It provides sensitivity in the nanomolar to low micromolar range making it suitable for precise measurements in vitro This kit is commonly applied in cell models including primary hepatocytes and macrophages as well as in animal studies for understanding lipid transport mechanisms It is also valuable in drug screening for identifying potential modulators of PLTP activity Experimental concentrations typically depend on the specific research design allowing for flexibility across varied experimental setups
Catalog No. 50-005-67796 Supplier Apexbio Technology LLC Supplier No. K2087100
Add to Cart
Add to Cart
PLTP Activity Fluorometric Assay Kit is utilized in biomedical research to quantify phospholipid transfer protein (PLTP) activity This kit is often used in studies focused on lipid metabolism and cardiovascular disease The assay operates through the detection of fluorescent signals generated when PLTP catalyzes the transfer of phospholipids It provides sensitivity in the nanomolar to low micromolar range making it suitable for precise measurements in vitro This kit is commonly applied in cell models including primary hepatocytes and macrophages as well as in animal studies for understanding lipid transport mechanisms It is also valuable in drug screening for identifying potential modulators of PLTP activity Experimental concentrations typically depend on the specific research design allowing for flexibility across varied experimental setups