Learn More
Waters Corp Plug, 1/4-28, Teflon, 2 per Package
Supplier:  Waters Corp 700009842
Encompass_Preferred
This plug is a replacement for 1/4-28 flat bottom ports. It is made from Teflon™ and is used to block unused ports.
  • Fitting type
  • Suitable for HPLC systems
  • Constructed from PTFE material
  • Packaged with 2 units per package
  • Replacement as required
Catalog No. 50-239-2662