Learn More
Labcon North America Plug Style Caps for Urine Centrifuge tubes, in Bags
Catalog No. 50101719
Encompass_Preferred
This item has a minimum qty of 2 per supplier requirements.
Cap, Plug style cap for Urine centrifuge tubes; 15mL; Labcon; Endotoxin Free (non-pyrogenic); Polyethylene USP Classs VI resin; Not Freezable; Not autoclavable; 500 per bag 2 bags per case; Made in USA
Catalog No. 50-101-719 Supplier Labcon North America Supplier No. 30558700109
This item has a minimum qty of 2 per supplier requirements.
Cap, Plug style cap for Urine centrifuge tubes; 15mL; Labcon; Endotoxin Free (non-pyrogenic); Polyethylene USP Classs VI resin; Not Freezable; Not autoclavable; 500 per bag 2 bags per case; Made in USA