Learn More
Grainger Plug 5-15P 15A 125VAC 1 Phase
Catalog No. 502765647
Encompass_Preferred
The Grainger Approved Plug is a 15 amp, 125 volt, NEMA 5-15P, 3-prong, grounded plug made of thermoplastic. It has a maximum temperature rating of 167°F and is UL listed. The plug measures 1-1/4"W x 2-1/8"L x 1-3/8"H overall. The prongs are 0.170" diameter and spaced 0.750" between centers. The plug body accepts 12-10 AWG wire. This general purpose plug can be used on extension cords, power tools, appliances, and more.
Catalog No. 50-276-5647 Supplier Grainger Supplier No. 16R088
The Grainger Approved Plug is a 15 amp, 125 volt, NEMA 5-15P, 3-prong, grounded plug made of thermoplastic. It has a maximum temperature rating of 167°F and is UL listed. The plug measures 1-1/4"W x 2-1/8"L x 1-3/8"H overall. The prongs are 0.170" diameter and spaced 0.750" between centers. The plug body accepts 12-10 AWG wire. This general purpose plug can be used on extension cords, power tools, appliances, and more.