Learn More
Grainger Plug 6-15P 15A 250VAC 1 Phase
Catalog No. 502765649
Encompass_Preferred
The Grainger 16R090 is a Westward brand 1/4-inch drive socket set. This 108-piece socket set includes both SAE and metric sockets ranging from 1/4 inch drive to 3/4 inch drive in shallow, deep, and universal depths. The set is made from chrome vanadium steel and comes packed in a sturdy plastic blow mold case for storage and portability. This socket set provides a wide assortment of the most common socket sizes needed for automotive and machinery repair and maintenance.
Catalog No. 50-276-5649 Supplier Grainger Supplier No. 16R090
The Grainger 16R090 is a Westward brand 1/4-inch drive socket set. This 108-piece socket set includes both SAE and metric sockets ranging from 1/4 inch drive to 3/4 inch drive in shallow, deep, and universal depths. The set is made from chrome vanadium steel and comes packed in a sturdy plastic blow mold case for storage and portability. This socket set provides a wide assortment of the most common socket sizes needed for automotive and machinery repair and maintenance.