Learn More
Labcon North America Plug Style Caps for 12x75mm Culture Tubes, Natural Color, in Bags
Catalog No. 50101614
Encompass_Preferred
Cap; Fluted Plug Style; Labcon; Plug cap for culture tubes. Fits popular 12x75mm culture tubes; Cap includes flutes to securely grip sides of tubes; Endotoxin Free (non-pyrogenic); Polyethylene USP Classs VI resin; Not autoclavable; Natural uncolored; 1000 per bag/3 bags per case; Made in USA
Catalog No. 50-101-614 Supplier Labcon North America Supplier No. 33028000009
Cap; Fluted Plug Style; Labcon; Plug cap for culture tubes. Fits popular 12x75mm culture tubes; Cap includes flutes to securely grip sides of tubes; Endotoxin Free (non-pyrogenic); Polyethylene USP Classs VI resin; Not autoclavable; Natural uncolored; 1000 per bag/3 bags per case; Made in USA